Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026479.4 | NP_001021650.1 | 6 - 7 | 1320 | 63 | |
GenBank mRNA | AJ307058.1 | - | 6 - 7 | 1320 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |