Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027281.1 | NP_001022452.1 | 5 - 6 | 1011 | 90 | |
NM_001027282.1 | NP_001022453.1 | 1 - 2 | 207 | 90 | ||
NM_001027283.1 | NP_001022454.1 | 1 - 2 | 207 | 90 | ||
GenBank mRNA | AF088285.1 | - | 7 - 8 | 1183 | 90 | |
AF088286.1 | - | 1 - 2 | 248 | 90 | ||
AF088287.1 | - | 1 - 2 | 248 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |