Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171089.7 | NP_741102.1 | 1 - 2 | 155 | 72 | |
GenBank mRNA | L16530.1 | - | 1 - 2 | 155 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |