Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065331.4 | NP_497732.1 | 12 - 13 | 2166 | 77 | |
NM_065332.4 | NP_497733.1 | 5 - 6 | 977 | 77 | ||
NM_171099.5 | NP_741112.1 | 5 - 6 | 810 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |