Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066082.5 | NP_498483.1 | 8 - 9 | 1621 | 74 | |
NM_066083.7 | NP_498484.1 | 8 - 9 | 1854 | 74 | ||
NM_171173.4 | NP_741201.1 | 11 - 12 | 1849 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |