Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exos-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372617.1 | NP_001360028.1 | 6 - 7 | 611 | 91 | |
NM_171186.6 | NP_741217.1 | 7 - 8 | 633 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |