Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jnk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026099.5 | NP_001021270.1 | 7 - 8 | 929 | 88 | |
NM_001383114.1 | 7 - 8 | 936 | 88 | |||
NR_131561.1 | 7 - 8 | 893 | 88 | |||
GenBank mRNA | AB024085.1 | - | 7 - 8 | 936 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |