Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mei-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059493.7 | NP_491894.1 | 3 - 4 | 574 | 68 | |
GenBank mRNA | AF248052.1 | - | 3 - 4 | 556 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |