Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ser-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059553.5 | NP_491954.1 | 5 - 6 | 1564 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |