Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sol-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059687.7 | NP_492088.3 | 4 - 5 | 541 | 163 | |
GenBank mRNA | AY387697.1 | - | 4 - 5 | 506 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |