Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: goa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059707.6 | NP_492108.1 | 3 - 4 | 553 | 125 | |
GenBank mRNA | AY008140.1 | - | 3 - 4 | 459 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |