Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nud-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059771.6 | NP_492172.1 | 4 - 5 | 1131 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |