Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026460.3 | NP_001021631.1 | 7 - 8 | 2587 | 75 | |
NM_001026461.4 | NP_001021632.1 | 7 - 8 | 2585 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |