Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cep-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026307.7 | NP_001021478.1 | 12 - 13 | 1954 | 80 | |
NM_001383463.1 | 11 - 12 | 1628 | 80 | |||
GenBank mRNA | AF440800.2 | - | 11 - 12 | 1634 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |