Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129002.4 | NP_001122474.1 | 7 - 8 | 825 | 88 | |
NM_059955.7 | NP_492356.1 | 7 - 8 | 814 | 88 | ||
GenBank mRNA | AF228692.1 | - | 7 - 8 | 836 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |