Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 11 - 12 | 1783 | 116 | |
NM_001026219.5 | NP_001021390.1 | 11 - 12 | 1734 | 116 | ||
NM_001128984.4 | NP_001122456.2 | 13 - 14 | 1950 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |