Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gfi-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381135.1 | NP_001366991.1 | 6 - 7 | 1535 | 82 | |
NM_001381136.2 | NP_001366793.1 | 6 - 7 | 1518 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |