Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oct-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026311.7 | NP_001021482.1 | 6 - 7 | 673 | 101 | |
NM_060216.6 | NP_492617.4 | 7 - 8 | 736 | 101 | ||
GenBank mRNA | AF110415.1 | - | 6 - 7 | 681 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |