Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025871.6 | NP_001021042.2 | 6 - 7 | 738 | 80 | |
NM_001025872.6 | NP_001021043.2 | 4 - 5 | 632 | 80 | ||
NM_001356801.3 | NP_001343721.1 | 5 - 6 | 720 | 80 | ||
GenBank mRNA | AY204173.1 | - | 5 - 6 | 729 | 80 | |
AY204174.1 | - | 4 - 5 | 654 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |