Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058831.7 | NP_491232.1 | 4 - 5 | 2161 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |