Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322672.3 | NP_001309486.1 | 4 - 5 | 438 | 69 | |
NM_001383080.1 | 8 - 9 | 989 | 69 | |||
NM_059174.8 | NP_491575.1 | 8 - 9 | 1183 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |