Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026404.7 | NP_001021575.1 | 3 - 4 | 626 | 104 | |
NM_059228.6 | NP_491629.1 | 3 - 4 | 620 | 104 | ||
NM_059229.7 | NP_491630.1 | 4 - 5 | 650 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |