Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313222.3 | NP_001300151.1 | 4 - 5 | 554 | 75 | |
NM_001313223.3 | NP_001300152.1 | 4 | 526 | 75 | ||
NM_171456.8 | NP_741536.1 | 4 - 5 | 560 | 75 | ||
NM_171928.6 | NP_741537.1 | 4 - 5 | 552 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |