Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rde-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171525.6 | NP_741611.1 | 10 - 11 | 2863 | 90 | |
GenBank mRNA | AF180730.1 | - | 10 - 11 | 2853 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |