Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269533.2 | NP_001256462.1 | 2 - 3 | 139 | 73 | |
NM_001269534.3 | NP_001256463.1 | 1 - 2 | 67 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |