Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375019.2 | NP_001361932.1 | 8 | 1265 | 96 | |
NM_001375020.2 | NP_001362134.1 | 7 - 8 | 1263 | 96 | ||
NM_001375021.2 | NP_001361933.1 | 10 - 11 | 2813 | 96 | ||
NM_001375022.2 | NP_001362133.1 | 10 - 11 | 2815 | 96 | ||
NM_001375024.2 | NP_001362136.1 | 9 - 10 | 2740 | 96 | ||
NM_001383564.1 | 10 - 11 | 2811 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |