Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368504.4 | NP_001355460.1 | 6 | 1097 | 74 | |
NM_001368505.3 | NP_001355457.1 | 10 - 11 | 1166 | 74 | ||
NM_001368507.2 | NP_001355459.1 | 10 - 11 | 1179 | 74 | ||
NM_001368511.3 | NP_001355478.1 | 10 - 11 | 1142 | 74 | ||
NM_001368515.4 | NP_001355479.1 | 10 | 1118 | 74 | ||
NM_001380971.2 | NP_001368710.1 | 10 - 11 | 1283 | 74 | ||
NM_001380972.1 | NP_001368712.1 | 11 - 12 | 1343 | 74 | ||
NM_001380973.1 | NP_001368715.1 | 11 - 12 | 1356 | 74 | ||
NM_001380974.1 | NP_001368713.1 | 10 - 11 | 1142 | 74 | ||
NM_001380975.1 | NP_001368716.1 | 10 - 11 | 1155 | 74 | ||
NM_001380976.1 | NP_001368714.1 | 10 - 11 | 1118 | 74 | ||
NM_001380977.1 | NP_001368717.1 | 10 - 11 | 1131 | 74 | ||
NM_171691.6 | NP_741797.1 | 11 - 12 | 1386 | 74 | ||
GenBank mRNA | U26735.1 | - | 11 - 12 | 1380 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |