Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029730.6 | NP_001024901.1 | 7 - 8 | 988 | 80 | |
NM_001375064.2 | NP_001362128.1 | 7 | 988 | 80 | ||
NM_001375065.2 | NP_001362126.1 | 7 | 988 | 80 | ||
NM_001375066.2 | NP_001362127.1 | 7 | 988 | 80 | ||
NM_001375067.2 | NP_001362125.1 | 7 | 988 | 80 | ||
NM_171720.7 | NP_741831.1 | 7 - 8 | 1009 | 80 | ||
GenBank mRNA | AF257062.1 | - | 7 - 8 | 988 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |