Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029616.6 | NP_001024787.1 | 7 - 8 | 1232 | 87 | |
NM_001029617.5 | NP_001024788.1 | 7 - 8 | 1109 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |