Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028998.4 | NP_001024169.1 | 6 - 7 | 1095 | 94 | |
NM_001028999.5 | NP_001024170.1 | 6 - 7 | 1446 | 94 | ||
NM_001029004.5 | NP_001024175.1 | 5 - 6 | 869 | 94 | ||
GenBank mRNA | AF427475.1 | - | 6 - 7 | 1117 | 94 | |
AF427477.1 | - | 6 - 7 | 1300 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |