Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029536.5 | NP_001024707.1 | 5 - 6 | 2020 | 107 | |
NM_001381134.1 | NP_001366990.1 | 5 - 6 | 2020 | 107 | ||
GenBank mRNA | AF200706.1 | - | 5 - 6 | 2027 | 107 | |
AF200707.1 | - | 5 - 6 | 2025 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |