Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065706.5 | NP_498107.2 | 5 - 6 | 546 | 128 | |
NM_181964.4 | NP_871693.1 | 5 - 6 | 568 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |