Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmp-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028960.5 | NP_001024131.1 | 6 - 7 | 804 | 62 | |
NM_072288.6 | NP_504689.1 | 5 - 6 | 667 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |