Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: marc-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060422.6 | NP_492823.2 | 6 - 7 | 2642 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Inventoried Probe spans exons |