Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060692.3 | NP_493093.1 | 8 - 9 | 1467 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |