Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060728.3 | NP_493129.3 | 6 - 7 | 1302 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |