Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sur-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026292.5 | NP_001021463.1 | 5 - 6 | 1133 | 132 | |
NM_061174.6 | NP_493575.2 | 5 - 6 | 1134 | 132 | ||
GenBank mRNA | U33051.1 | - | 5 - 6 | 1135 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |