Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubxn-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027010.5 | NP_001022181.2 | 8 - 9 | 1208 | 62 | |
NM_001313618.3 | NP_001300547.1 | 5 - 6 | 833 | 62 | ||
NM_001313620.3 | NP_001300549.1 | 5 - 6 | 848 | 62 | ||
NM_001383772.1 | 10 - 11 | 1336 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |