Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061534.4 | NP_493935.3 | 7 - 8 | 1267 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |