Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cogc-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061565.6 | NP_493966.1 | 4 - 5 | 1115 | 122 | |
NM_061566.7 | NP_493967.1 | 3 - 4 | 780 | 122 |
Gene Symbol: Y51H7C.41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381330.2 | NP_001368658.1 | 6 - 7 | 1663 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |