Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apd-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062169.6 | NP_494570.1 | 3 - 4 | 299 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |