Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062406.6 | NP_494807.1 | 3 - 4 | 406 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |