Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062406.6 | NP_494807.1 | 5 - 6 | 954 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |