Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383859.1 | 8 - 9 | 1946 | 154 | ||
NR_165322.1 | 8 - 9 | 1892 | 154 | |||
NR_165323.1 | 8 - 9 | 1892 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |