Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026958.5 | NP_001022129.1 | 6 - 7 | 1126 | 110 | |
GenBank mRNA | L15302.1 | - | 6 - 7 | 1148 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |