Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ltd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063296.6 | NP_495697.2 | 2 - 3 | 359 | 109 | |
GenBank mRNA | AF489839.1 | - | 2 - 3 | 381 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |