Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322749.3 | NP_001309578.1 | 9 | 1471 | 81 | |
NM_001322750.3 | NP_001309579.1 | 6 - 7 | 1053 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Amplicon spans exons and probe does not span exons |