Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026880.5 | NP_001022051.1 | 5 - 6 | 1741 | 87 | |
NM_001383916.1 | 5 - 6 | 1733 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |