Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aqp-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063704.6 | NP_496105.1 | 4 - 5 | 615 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |