Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdt-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063815.5 | NP_496216.1 | 2 - 3 | 332 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |